Wearwell™ Tile-Top Antimicrobial Mats
Permanent antimicrobial surface destroys microorganisms on contact
$147.87 - $779.77
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Price | Quantity | ||||
19-151-1212
|
Wearwell™
420.12X3X5AMBK |
Black | 152.4 x 91.4 cm (5 x 3 ft.) |
Each for $367.92
|
|
||||
19-151-1213
|
Wearwell™
420.12X3X5AMBL |
Blue | 152.4 x 91.4 cm (5 x 3 ft.) |
Each for $332.81
|
|
||||
19-151-1214
|
Wearwell™
420.12X3X5AMGY |
Gray | 152.4 x 91.4 cm (5 x 3 ft.) |
Each for $332.81
|
|
||||
19-151-1210
|
Wearwell™
420.12X2X3AMGY |
Gray | 91.4 x 61 cm (3 x 2 ft.) |
Each for $167.10
|
|
||||
19-151-1208
|
Wearwell™
420.12X2X3AMBK |
Black | 91.4 x 61 cm (3 x 2 ft.) |
Each for $151.28
|
|
||||
19-151-1211
|
Wearwell™
420.12X2X3AMCH |
Charcoal Gray | 91.4 x 61 cm (3 x 2 ft.) |
Each for $166.64
|
|
||||
19-151-1215
|
Wearwell™
420.12X3X5AMCH |
Charcoal Gray | 152.4 x 91.4 cm (5 x 3 ft.) |
Each for $401.21
|
|
||||
19-151-180
|
Wearwell™
420.12X3X10AMBK |
Black | 91 x 304 cm (3 x 10 ft.) |
Each for $779.77
|
|
||||
19-151-182
|
Wearwell™
420.12X3X10AMCH |
Charcoal Gray | 91 x 304 cm (3 x 10 ft.) |
Each for $779.77
|
|
||||
19-151-181
|
Wearwell™
420.12X3X10AMBL |
Blue | 91 x 304 cm (3 x 10 ft.) |
Each for $779.77
|
|
||||
19-003-153B
|
Wearwell™
420.12X2X3AMBL |
Blue | 91.4 x 61 cm (3 x 2 ft.) |
Each for $147.87
|
|
||||
Description
Permanent antimicrobial surface destroys microorganisms on contact
- Ideal for use in all dry work areas from packaging, banks, labs, office and behind or in front of counters
- Unique antimicrobial Nitricell™ sponge base offers long life and resilience
- 0.5 in. (1.3cm) sponge base for extra comfort
- Extra wear layer for abrasion and chemical resistance
Specifications
0.5 in. |