Wearwell™ UltraSoft Tile-Top Antimicrobial Mats
100% antimicrobial mat
Supplier:  Wearwell™ 419.78X3X5AMBL
 View more versions of this product
Catalog No. 19-151-1205
Explore available promotions
Description
Description
  • Permanent antimicrobial nonporous vinyl surface with micro-pure™ nitrile composite sponge base
  • 0.875 in. (2.22cm) double thick sponge base for extra comfort
  • Destroys microorganisms on contact
  • Extra wear layer for abrasion and chemical resistance
Not recommended for use under chairs, stools and casters, or with spiked heels.
Order Info
Mats may also be custom ordered in virtually any size or dimension to fit very large and unique spaces. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Specifications
152.4 x 91.4 cm (5 x 3 ft.)
Blue
Nitricell, PVC