Wearwell™ Upfront Scraper Entrance Mat
For use at heavily trafficked outdoor entrances, water or drink fountains
Supplier:  Wearwell™ 224.516X3X5GRBK
 View more versions of this product
Catalog No. 19-150-0090
Explore available promotions
Description
Description
  • Elevates employees above the area where water spill-over will collect
  • Keeps water off floors
  • Pliable and durable rubber compound is highly resistant to harsh outside elements
  • Low profile accommodates tight doorways
  • Easily cleaned, simply mop it or shake it off
  • Color: Black
  • Use the slotted version as an outdoor entrance mat
Specifications
Specifications
91 x 152 cm
Black
Unslotted