missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 4–20%, 1.0 mm, WedgeWell™ format DFS Item

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.

$45.32 - $184.00

Spécifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 4 to 20%
Gel Size Mini
GelThickness 1.0 mm
Gel Type Tris-Glycine Plus
voir les spécifications

Produits 4
Numéro de catalogue Numéro du manufacturier. Wells Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Wells Quantity Prix Quantité  
XP04200PK2 DFS Item
Afficher les documents
Invitrogen™
XP04200PK2
10-well 2 gels (1 box)
paquet de 2 chaque for $45.32
 
XP04200BOX DFS Item
Afficher les documents
Invitrogen™
XP04200BOX
10-well 10 gels (1 box)
paquet for $184.00
 
XP04202BOX DFS Item
Afficher les documents
Invitrogen™
XP04202BOX
12-well 10 gels (1 box)
paquet for $184.00
 
XP04205BOX DFS Item
Afficher les documents
Invitrogen™
XP04205BOX
15-well 10 gels (1 box)
paquet for $184.00
 
Description

Description

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.

Features of Novex Tris-Glycine gels, WedgeWell format, include:
Improved shelf life—store gels for up to 7 months at 4°C
Diversity—use for native and denaturing protein assays
Wedge-shaped wells—easily load up to two times more sample volume
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.

Run your proteins in native or denatured form
Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

Convenient to use
Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000). Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

Spécifications

Spécifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Tris-Glycine Plus
20 to 200 kDa
Molecular Weight
Wet Ice
8 cm
4 to 20%
1.0 mm
Denaturing, Native
Denaturing, Native
Novex™, WedgeWell™
Store at 2°C to 8°C. Do not freeze.
8 cm
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'