METTLER TOLEDO™ Weighing Table for Micro Analytical Balance
Professional Weighing Table
Supplier:  METTLER TOLEDO™ 11138042
Catalog No. 13940083
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Speeds up weighing process
  • With two-tables-in-one, the balance is placed separately from working area to ensure vibration free operation and fast stabilization times
  • Ergonomically designed
  • Height adjustable outer table relaxes users' neck and back muscles
  • Elbow cushions protect arms during repetitive activities
  • Accommodates a monitor and keyboard and provides neat cable management.
Specifications
Specifications
130 kg
784 mm
760 mm
Height
Weighing Table
803 mm
XPE26/56 micro balances