Welch™ DuoSeal™ Pump Oil
For use in DuoSeal High-Vacuum Pumps
Supplier:  Welch™ 1407K15
 View more versions of this product
Catalog No. 01-104B
Explore available promotions
Description
Description
For use in DuoSeal High-Vacuum Pumps
For use in DuoSeal High-Vacuum Pumps. One gallon.Specially fractionate oil has properties enabling highest performance for DuoSeal pumps
  • Low vapor Pressure
  • High Viscosity
Specifications
Specifications
Vaccum Pump Oil
DuoSeal™ Belt-driven pumps
1 gal.
Provides the lubrication that helps enable renowned DuoSeal™ pump durability and ensure the highest vacuum performance
1 Ea.
3.78 L
Product Suggestions
Product Suggestions