Welch™ Premium Pump Oil
Supplier:  Welch™ 8995P15
 View more versions of this product
Catalog No. 01-131-27
Explore available promotions
Description
Description
Specifications
Specifications
Premium Pump Oil
Welch Premium Vacuum Pump
1 gal.
Resists breakdown in the high RPM operation environment of direct drive pumps and enables direct drive pumps to maintain the highest vacuum performance over time
1 Ea.
3.78 L