Learn More
3M™ Welding Hard Hat
3M™ Welding Hard Hat with welding shield and wide-view faceshield for use with Speedglas™ 9100 Series Auto-Darkening Filter.
Supplier: 3M™ 7000052730
Description
- For use with Speedglas™ 9100 Series Auto-Darkening Filter
- 3M Catalog# L-705SG
Specifications
| ANSI Z89.1 Type II, Class G | |
| L-705SG | |
| 1 Ea. | |
| Hardhat with Faceshield and Welding Shield | |
| Not Ventilated |
| Green | |
| Headgear | |
| Standard | |
| Clear Visor and Auto-Darkening Welding Lens |