missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Western Stripping Buffer

Western stripping buffer simplifies the western blot process by removing primary and secondary antibodies from membranes, thus enabling the reuse of membranes for reprobing with different primary antibodies.

Supplier:  MP Biomedicals™ 08W00019

Catalog No. ICN08W00019


Explore available promotions

Description

Description

MP Bio™'s Western Stripping Buffer adopts a neutral washing formula for removing bound primary and secondary antibodies from membranes, without removing or damaging the immobilized antigen. It eliminates time and cost waste by allowing the reuse of nitrocellulose or PVDF blots to detect different targets with different primary antibodies, without the need to re-run gels and transfer.

  • pH Neutral Stripping Buffer
  • Strip antibodies from membranes
  • Re-probe membranes after stripping
Specifications

Specifications

Western Blot Stripping Buffer
Liquid
Western Blot
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.