missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Whatman™ 3 Chr Chromatography Paper

Pure cellulose produced from cotton linters with no additives

Supplier:  Cytiva 3003917

Catalog No. 05-716-4B


Explore available promotions

Description

Description

  • Typical thickness: 0.36mm
  • Typical linear flowrate: 130mm/30 min.
  • General applications with medium/heavy solute loadings

Specifications

Specifications

46 x 57 cm (18.1 x 22.4 in.)
Grade 3 Chr cellulose chromatography sheets is a 0.36 mm paper for general applications with medium/heavy solute loadings.
0.36 mm
46 cm
Cellulose
57 cm
18.1 in.
Sheet
For General AppliCatIons with Medium/Heavy Solute Loadings, Frequently Used for Separation of Inorganic Compounds and for Electrophoresis
Grade 3 Chr
130 mm/30 min.
Videos
SDS
Documents

Documents

Promotions

Promotions