Learn More
Cytiva Whatman™ 903 Protein Saver Card
For research use only
Supplier: Cytiva 10534612
Description
For research use only
- Each card has five half-inch circles printed on the sample collection area
- Each circle holds 75 to 80 μL of sample
- Wraparound cover has spaces for name and date of collection and is imprinted with universal biohazard symbol in accordance with USPS regulations
- Fits into Whatman foil barrier zipper bags for storage
Specifications
| Five 0.5 in. circles holding 75 to 80 μL sample, wraparound cover with spaces for name and date of collection, imprinted with universal biohazard symbol | |
| 100/Pk. | |
| Protein Saver Card |