Cytiva Whatman™ Antibiotic Assay Discs
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Supplier:  Cytiva 2017013
 View more versions of this product
Catalog No. 05-712
Explore available promotions
Description
Description
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
  • For use with Kirby-Bauer method of antibiotic testing
  • Allows rational and selective chemotherapy
Specifications
Specifications
Antibiotic Assay Discs
1000 Discs/Pk.
13 mm
Infectious Disease, Antibiotic Susceptibility Sensitivity
1000 Pack