Cytiva Whatman™ CyDye™ Cy3 Mono NHS Ester
For use in a variety of applications at biologically relevant pHs
Supplier:  Cytiva 25900472
Catalog No. 09-928-079
Explore available promotions
Description
Description
  • Versatile fluorophores that can tolerate a pH range of 3 to 10
  • Dyes are DMSO tolerant and photostable to enable transfer from storage to assay without loss of performance
  • Aqueous solubility eliminates the need for organic solvents in the assay buffers
  • Fluors are thoroughly QC tested to ensure high levels of chromophore and reactive dye content
Specifications
Specifications
150 μg
150 μg
3 to 10
CyDye™
Suitable for Targeted, Precise Labeling of Amine Groups, Proteins, and Oligonucleotides
Fluorescent Labeling Reagent
Red