Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper
17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
Supplier:  Cytiva 3017917
 View more versions of this product
Catalog No. 09924555
Explore available promotions
Description
Description
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
Specifications
Specifications
46 x 57 cm (18.1 x 22.4 in.)
22.4 in.
Sheet
Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Grade 17 Chr
190 mm/30 min.
18.1 in.
17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
0.92 mm
46 cm
Cellulose
57 cm