Cytiva Whatman™ Grade 2668 Chr Cellulose Chromatography Paper
2668 Chr cellulose chromatography sheets is a 0.9 mm paper for separation relatively large molecules by electrophoresis.
Supplier:  Cytiva 10382461
Catalog No. 09-800-949
Explore available promotions
Description
Description
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
Specifications
Specifications
58 x 60 cm (22.8 x 23.6 in.)
2668 Chr cellulose chromatography sheets is a 0.9 mm paper for separation relatively large molecules by electrophoresis.
Grade 2668 Chr