Cytiva Whatman™ Nuclepore Hydrophilic Membrane
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
$118.35 - $846.00
Specifications
Material | Polycarbonate |
---|---|
For Use With (Application) | Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses |
Type | Nuclepore hydrophilic membrane |
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) | Price | Quantity | ||||
18400175
|
Cytiva
10418701 |
1 μm | 13 mm |
Pack of 100 Each for $118.35
|
|
||||
18400180
|
Cytiva
10418704 |
1 μm | 19 mm |
Pack of 100 Each for $129.74
|
|
||||
18400184
|
Cytiva
10418712 |
1 μm | 47 mm |
Pack of 100 Each for $263.00
|
|
||||
09920011
|
Cytiva
10417506 |
8 μm | 25 mm |
Pack of 100 Each for $157.37
|
|
||||
09920018
|
Cytiva
10417512 |
8 μm | 47 mm |
Pack of 100 Each for $273.00
|
|
||||
18400173
|
Cytiva
10418401 |
10 μm | 13 mm |
Pack of 100 Each for $171.89
|
|
||||
18400182
|
Cytiva
10418406 |
10 μm | 25 mm |
Pack of 100 Each for $152.08
|
|
||||
18400179
|
Cytiva
10418412 |
10 μm | 47 mm |
Pack of 100 Each for $263.00
|
|
||||
18400181
|
Cytiva
10418501 |
12 μm | 13 mm |
Pack of 100 Each for $125.28
|
|
||||
18400176
|
Cytiva
10418506 |
12 μm | 25 mm |
Pack of 100 Each for $136.10
|
|
||||
18400177
|
Cytiva
10418512 |
12 μm | 47 mm |
Pack of 100 Each for $237.00
|
|
||||
18400171
|
Cytiva
10418718 |
1 μm | 90 mm |
Pack of 25 Each for $308.00
|
|
||||
18400172
|
Cytiva
10418739 |
1 μm | 293 mm |
Pack of 25 Each for $846.00
|
|
||||
18400174
|
Cytiva
10418301 |
3 μm | 13 mm |
Pack of 100 Each for $118.35
|
|
||||
18400178
|
Cytiva
10418806 |
2 μm | 25 mm |
Pack of 100 Each for $152.08
|
|
||||
18400187
|
Cytiva
10418812 |
2 μm | 47 mm |
Pack of 100 Each for $263.00
|
|
||||
18400186
|
Cytiva
10418306 |
3 μm | 25 mm |
Pack of 100 Each for $152.08
|
|
||||
18400185
|
Cytiva
10418312 |
3 μm | 47 mm |
Pack of 100 Each for $263.00
|
|
||||
Description
- Track-etched for highly defined pore size and high pore density
- Treated with PVP to make it hydrophilic
- Low extractables minimizing filtration sample contamination risk
- High chemical resistance
- Good thermal stability
- Low, consistent ash
- Low tare weights
- Smooth, flat membrane surface for high particle visibility
Applications:
- Bioassays
- Cell biology
- Environmental analysis
- Microscopy
- Cytology
- Parasitology
- Certain air and oceanography analyses
Specifications
Polycarbonate | |
Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses | |
Nuclepore hydrophilic membrane |