missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Whatman™ Nylon Membrane Circle

For filtering aqueous solutions and most organic solvents

Supplier:  Cytiva 7402002

Catalog No. 09-910-32A


Explore available promotions

Description

Description

For filtering aqueous solutions and most organic solvents

  • Hydrophilic- eliminating need for wetting agents
  • For use with a wide range of biological preparations
  • Flexible, durable and tear resistant
  • Can be autoclaved at 135°C

Specifications

Specifications

Autoclavable
Nylon
For filtering aqueous solutions and most organic solvents
Filtration Membrane Disc
Membrane Filter
0.2 μm
Hydrophilic
25 mm
Videos
SDS
Documents

Documents

Promotions

Promotions