missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Whatman™ Plastic Zipper Seal Storage Bag

Zipper storage bag for 903 Protein Saver Cards

Supplier:  Cytiva 10548232

Catalog No. 0980016


Explore available promotions

Description

Description

  • Accommodates cards up to W x H: 95 x 130 mm
  • W x H: 4 x 6 in. (102 x 152 mm)
  • Clear flexible plastic
  • Pack of 100
Specifications

Specifications

Zip-seal Storage Bag
Clear
Plastic
100
102 mm
152 mm
Zip Seal
Storing and archiving Whatman Protein Saver card samples.
100
4 in.
6 in.
Videos
SDS
Documents

Documents

Promotions

Promotions