Cytiva Whatman™ Polycap Disposable Capsules, 36 AS
Provides highly effective filtration area in a small size. Whatman™ Polycap Disposable AS Capsules, AS 36 filters large volumes and difficult samples with ease.
Supplier:  Cytiva 67093602
 View more versions of this product
Catalog No. 09-743-94
Explore available promotions
Description
Description
Recommended for filtering aqueous solutions. Combines a glass microfiber (GMF) prefilter and a nylon membrane, prolonging the life of the filter and allowing larger volumes and difficult samples to be filtered easily.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 μm) life and higher filtration efficiency.
  • Nylon membrane layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates.
  • Housing thermally fused (no glues, adhesives, or extraneous materials).
  • Integrity-testable by bubble point, pressure decay, or forward flow methods.
  • Provides highly effective filtration area in a small size.
  • Autoclavable; some presterilized with gamma irradiation.
  • Manufactured in clean room facilities under ISO Quality Systems.
Specifications
Specifications
Autoclavable
Nylon
Sterile
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
MNPT inlet
SB outlet
Polycap AS 36 Capsule Filter, sterile, 0.2 μm, MNPT inlet and SB outlet (1 pc)
0.2 μm
400 cm2
Polypropylene (PP)
4.1 bar (60 psi)