missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ SG81 Grade Chromatography Paper
SG81 ion exchange sheets is a 0.27 mm combining cellulose and large pore silica gel for separations in which both partition and adsorption are important.
Supplier: Cytiva 3668915
Description
- Simultaneous development of multiple samples on the same sheet under identical conditions.
- Sequential development of the same samples with different solvents and/or different concentrations of the same solvent.
- Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution.
- Used in enzyme assays to separate product from reactant by charge or polarity.
Specifications
18.1 in. | |
0.27 mm | |
46 cm | |
Cellulose, Large Pore Silica | |
57 cm |
Sheet | |
For Enzyme Assays to Separate Product From Reactant by Charge or Polarity | |
SG81 | |
110 mm/30 min. |