DWK Life Sciences Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached
For optimal performance in the laboratory. Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached offer low permeability to solvents and minimal background counts.
Supplier:  DWK Life Sciences 986734
 View more versions of this product
Catalog No. 03-341-73C
Explore available promotions
Description
Description
  • Clarity of glass with the safety of plastic
  • Vial can be incinerated; no harmful gas is generated
  • Vials offer low permeability to solvents and minimal background counts
Specifications
Specifications
Linerless
White
PET
Offer low permeability to solvents and minimal background counts, vials can be safely incinerated, no harmful gas is generated
Tray
Polyethylene
61 mm
1.06 in.
27 mm
20 mL
2.4 in.
0.67 oz.
Screw Cap
Videos
SDS
Documents
Documents
Accessories