DWK Life Sciences Wheaton™ Fritted Glass Support for 47mm Filtration Assemblies with No. 8 Stopper Connections
Recommended for general filtration. Wheaton™ Fritted Glass Support for 47mm Filtration Assemblies with No. 8 Stopper Connections is manufactured from Wheaton™ 33 low extractable borosilicate glass.
Supplier:  DWK Life Sciences 419352
Catalog No. 14-209-114
Explore available promotions
Description
Description
  • Glass conforms to ASTM E 438 Type I, Class A requirements
  • Includes a coarse porosity (40-60μm) fritted glass support base
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class A
Glass
Fritted Support Base
Filtration Assembly (47 mm)
1 Case