missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Fritted Glass Support Base for use with Wheaton 47mm Filtration Assembly

Replacement Glass Support Base with 40/35 [ST] Joint Connection for use with DWK Life Sciences Wheaton 47mm Glass Filter Holder Assembly (419380).

Supplier:  DWK Life Sciences 419382

Catalog No. 14-209-126


Explore available promotions

Description

Description

  • Fritted glass support base for Wheaton 47mm Vacuum Filter Holder Assemblies
  • Manufactured from Wheaton 33 low extractable borosilicate glass which conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
  • Coarse porosity (40 to 60μm)
Specifications

Specifications

USP Type I, ASTM E 438 Type I, Class A
Glass
Fritted Support Base
47mm filtration assembly
Case of 1
SDS
Documents

Documents

Promotions

Promotions