Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
DWK Life Sciences Wheaton™ Allihn Condensers For Soxhlet Extraction Apparatus
Used for continuous extraction of analytes from a solid into an organic solvent
Supplier:  DWK Life Sciences 334025
 Afficher plus de versions de ce produit
Numéro de catalogue. 02-912-576
Découvrez plus d'offres spéciales
Description
Description
  • Wetted parts are manufactured from WHEATON 33 low extractable borosilicate glass
  • How it works:
    • The flask containing the solvent is heated
    • Vapors rise in the larger outside tube
    • Vapors enter the water–cooled condenser and liquate
    • When the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask
    Compliance
    WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
    Spécifications
    Spécifications
    Larger outside tube and bent tube
    Used for continuous extraction of analytes from a solid into an organic solvent
    Borosilicate Glass
    USP Type I and ASTM E438 Type I, Class A
    55/50
    Allihn Condenser
    Soxhlet extraction apparatus
    Videos
    FDS
    Documents
    Documents
    Offres spéciales
    Offres spéciales