DWK Life Sciences Wheaton™ Amber Wide-Mouth Packers without Closures
Amber glass for storage of light-sensitive powders
$244.21 - $711.38
Specifications
Color Amber
Closure Type Screw Cap
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-912-206
View Documents Promotion Details
DWK Life Sciences
W216960
30 mL
Case of 432 Each for $711.38
02-912-207
View Documents Promotion Details
DWK Life Sciences
W216961
60 mL
Case of 216 Each for $470.68
02-912-208
View Documents Promotion Details
DWK Life Sciences
W216962
125 mL
Case of 180 Each for $475.65
02-912-209
View Documents Promotion Details
DWK Life Sciences
W216963
250 mL
Case of 84 Each for $244.21
02-912-210
View Documents Promotion Details
DWK Life Sciences
W216964
500 mL
Case of 60 Each for $268.50
02-912-211
View Documents Promotion Details
DWK Life Sciences
W216965
1000 mL
Case of 36 Each for $251.32
Description
Description
  • USP Type III soda-lime glass
  • Allows for easy removal of contents
Specifications
Specifications
Amber
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'