DWK Life Sciences Wheaton™ BOD Bottles
Available with either glass robotic or pennyhead stoppers
$1106.06 - $4499.71
Specifications
For Use With (Application) | Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action |
---|---|
Marking Spot | Yes |
Material | Borosilicate Glass |
Catalog Number | Mfr. No. | Volume (Metric) | No. Sequence | Stopper | Barcode | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | No. Sequence | Stopper | Barcode | Price | Quantity | ||
02-912-547
|
DWK Life Sciences
22749499 |
60 mL | Specials | Robotic | Yes |
Case of 36 Each for $4,499.71
|
|
||
02-912-549
|
DWK Life Sciences
22749713G |
300 mL | 289 to 312 | Pennyhead | Yes |
Case of 24 Each for $1,294.91
|
|
||
02-926-11
|
DWK Life Sciences
22749703G |
300 mL | 49 to 72 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-12
|
DWK Life Sciences
22749400 |
60 mL | Unnumbered | Robotic | No |
Case of 36 Each for $3,228.65
|
|
||
02-926-13
|
DWK Life Sciences
22749401 |
60 mL | 1 to 36 | Robotic | Yes |
Case of 36 Each for $3,447.91
|
|
||
02-926-25
|
DWK Life Sciences
22749704G |
300 mL | 73 to 96 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-26
|
DWK Life Sciences
22749700 |
300 mL | Unnumbered | Robotic | Yes |
Case of 24 Each for $1,106.06
|
|
||
02-926-27
|
DWK Life Sciences
22749701 |
300 mL | 1 to 24 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-28
|
DWK Life Sciences
22749702 |
300 mL | 25 to 48 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-29
|
DWK Life Sciences
22749703 |
300 mL | 49 to 72 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-30
|
DWK Life Sciences
22749704 |
300 mL | 73 to 96 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-32
|
DWK Life Sciences
22749706 |
300 mL | 121 to 144 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-31
|
DWK Life Sciences
22749705 |
300 mL | 97 to 120 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-33
|
DWK Life Sciences
22749707 |
300 mL | 145 to 168 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-34
|
DWK Life Sciences
22749708 |
300 mL | 169 to 192 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-35
|
DWK Life Sciences
22749709 |
300 mL | 193 to 216 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-36
|
DWK Life Sciences
22749710 |
300 mL | 217 to 240 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-37
|
DWK Life Sciences
22749711 |
300 mL | 241 to 264 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-39
|
DWK Life Sciences
22749713 |
300 mL | 289 to 312 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-38
|
DWK Life Sciences
22749712 |
300 mL | 265 to 288 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-40
|
DWK Life Sciences
22749714 |
300 mL | 313 to 336 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-41
|
DWK Life Sciences
22749715 |
300 mL | 337 to 360 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-42
|
DWK Life Sciences
22749716 |
300 mL | 361 to 384 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-43
|
DWK Life Sciences
22749717 |
300 mL | 385 to 408 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-44
|
DWK Life Sciences
22749718 |
300 mL | 409 to 432 | Robotic | Yes |
Case of 24 Each for $1,157.71
|
|
||
02-926-8
|
DWK Life Sciences
22749701G |
300 mL | 1 to 24 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-7A
|
DWK Life Sciences
22749700G |
300 mL | Unnumbered | Pennyhead | Yes |
Case of 24 Each for $1,128.67
|
|
||
02-926-9
|
DWK Life Sciences
22749702G |
300 mL | 25 to 48 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-90
|
DWK Life Sciences
22749705G |
300 mL | 97 to 120 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-91
|
DWK Life Sciences
22749706G |
300 mL | 121 to 144 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-92
|
DWK Life Sciences
22749707G |
300 mL | 145 to 168 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-93
|
DWK Life Sciences
22749708G |
300 mL | 169 to 192 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-94
|
DWK Life Sciences
22749709G |
300 mL | 193 to 216 | Pennyhead | Yes |
Case of 24 Each for $1,275.45
|
|
||
02-926-95
|
DWK Life Sciences
22749710G |
300 mL | 217 to 240 | Pennyhead | Yes |
Case of 24 Each for $1,181.30
|
|
||
02-926-96
|
DWK Life Sciences
22749711G |
300 mL | 241 to 264 | Pennyhead | Yes |
Case of 24 Each for $1,115.48
|
|
||
Description
Available with either glass robotic or pennyhead stoppers
Bottles
- Borosilicate glass, 60 or 300mL, numbered or unnumbered
- With permanent marking areas; numbered versions are also bar-coded
- Specially designed shoulder radius sweeps all air from inside the bottle during filling
- 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
- Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
- Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
- Stopper joint is compatible with the probes of the leading BOD and DO meters
- Glass robotic stoppers are compatible with most robotic systems
Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.
Order Info
Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)
Specifications
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action | |
Yes | |
Borosilicate Glass |