DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Provide maximum surface between culture and glass flask
$240.81 - $1204.92
Specifications
Closure Material | Silicone Rubber |
---|---|
Surface Coating | PTFE Resin |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | ||||
06-416-2B
|
DWK Life Sciences
356875 |
50 mL | Flask, impeller and caps |
Case for $355.08
|
|
||||
06-416-2D
|
DWK Life Sciences
356879 |
250 mL | Flask, impeller and caps |
Each for $481.66
|
|
||||
06-416-2A
|
DWK Life Sciences
356873 |
25 mL | Flask, impeller and caps |
Case for $358.37
|
|
||||
06-416-2F
|
DWK Life Sciences
356887 |
3000 mL | Flask, impeller and caps |
Case for $1,204.92
|
|
||||
06-416-2J
|
DWK Life Sciences
DWK356884 |
1000 mL | Flask, impeller and caps |
Each for $743.75
|
|
||||
06-416-2C
|
DWK Life Sciences
356876 |
125 mL | Flask, impeller and caps |
Case for $438.83
|
|
||||
06-416-2E
|
DWK Life Sciences
356882 |
500 mL | Flask, impeller and caps |
Case for $637.52
|
|
||||
06-416-7B
|
DWK Life Sciences
356915 |
50 mL | Flask only |
Each for $240.81
|
|
||||
Description
Provide maximum surface between culture and glass flask
- PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
- Silicone rubber-lined cap
- Autoclavable
Specifications
Silicone Rubber | |
PTFE Resin | |
Autoclavable |