DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
Supplier:  DWK Life Sciences 224884
 View more versions of this product
Catalog No. 06-408C
Explore available promotions
Description
Description
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Cap and Cap Liners
    • Solid top, black phenolic caps
    • 14B rubber-lined black phenolic screw cap for aqueous samples
    • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Specifications
Specifications
14B Rubber
17 mm
8 mL
Screw
Wheaton
Phenolic
Screw Cap
ASTM Type I, Class A, USP Type I
15-425
Clear
Borosilicate Glass (Low Extractable)
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
2.2 dr.
8 mL
Autoclavable
63 mm
Vial
M-T File™ and Vial Rack
Product Suggestions
Product Suggestions