missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Ideal for soil sampling and environmental applications
$63.74 - $713.09
Specifications
Color | Clear |
---|---|
Material | Glass |
Closure Color | White |
Closure Liner | Poly/Vinyl |
Closure Material | Polypropylene |
Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
02-911-417
|
DWK Life Sciences
W216914 |
60 mL | Valu-bulk pack |
Case of 144 Each for $713.09
|
|
||||
02-911-406
|
DWK Life Sciences
W216903 |
60 mL | Convenience Pk. |
Case of 24 Each for $63.74
|
|
||||
02-911-407
|
DWK Life Sciences
W216904 |
125 mL | Convenience pack |
Case of 24 Each for $71.71
|
|
||||
02-911-408
|
DWK Life Sciences
W216905 |
250 mL | Convenience pack |
Case of 12 Each for $76.53
|
|
||||
02-911-409
|
DWK Life Sciences
W216906 |
500 mL | Convenience pack |
Case of 12 Each for $100.53
|
|
||||
02-911-410
|
DWK Life Sciences
W216907 |
1000 mL | Convenience pack |
Case of 12 Each for $114.35
|
|
||||
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
Clear-USP Type III soda-lime glass
Specifications
Clear | |
White | |
Polypropylene |
Glass | |
Poly/Vinyl | |
Screw Cap |