missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Ideal for soil sampling and environmental applications
Supplier: DWK Life Sciences W216909
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Walls allow for complete removal of contents
- Wide-mouth design is great for large solid samples
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear USP Type III soda-lime glass
Specifications
4 oz., 125 mL | |
4 oz. | |
2.36 in. | |
White | |
Yes | |
Round | |
Straight-sided Jar | |
PTFE | |
58-400 mm | |
Screw Cap |
125 mL | |
Clear | |
Soda Lime Glass | |
USP Type III | |
Wide | |
68 mm | |
2.6 in. | |
Polypropylene | |
60 mm | |
Yes |