missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Clear Vials for Environmental Analysis, with Caps Attached

Clear Vial with Open Top White Polypropylene PTFE Faced Silicone Lined Cap

$373.58 - $373.58

Specifications

Material (Liner) PTFE/Silicone
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
For Use With (Application) Environmental analysis
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Volume (English) Price Quantity  
02-912-517
View Documents Promotion Details
DWK Life Sciences
W224609
20 mL 60 mm 5 dr.
Case of 72 Each for $373.58
 
Description

Description

  • Septa of 5 mils of PTFE facing 120 mils of silicone is bonded to cap
Specifications

Specifications

PTFE/Silicone
28 mm
Environmental analysis
Polypropylene
Vial
Clear
Borosilicate Glass
Screw
Screw Cap (Open-top)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'