missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components

Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)

$74.15 - $392.56

Specifications

For Use With (Equipment) Cyanide distillation kit
View More Specs

Products 6
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
09-108-1
View Documents Promotion Details
DWK Life Sciences
377161
Absorber Jacket
Each for $123.24
 
09-108-4
View Documents Promotion Details
DWK Life Sciences
377164
Cold Finger Jacket
Each for $331.91
 
09-108-3
View Documents Promotion Details
DWK Life Sciences
377163
Cold Finger Condenser
Each for $250.09
 
09-108-5
View Documents Promotion Details
DWK Life Sciences
377165
Distilling Flask
Each for $299.76
 
09-108-2
View Documents Promotion Details
DWK Life Sciences
377162
Absorber Top
Each for $392.56
 
09-108-6
View Documents Promotion Details
DWK Life Sciences
377166
Inlet Tube
Each for $74.15
 
Description

Description

Compliance

For use according to ASTM D 2036

Specifications

Specifications

Cyanide distillation kit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'