DWK Life Sciences Wheaton™ Complete Cyanide Distillation Kit
Removes interference substances in cyanide quantitative analyses according to ASTM™ D 2036
Supplier: DWK Life Sciences 377160
Includes: Absorber jacket, absorber top, cold finger condenser, cold finger jacket, 1000mL distilling flask, and inlet tube
Description
Sample reacts with HS2O4 and MgCl2 to convert any cyanide to hydrocyanic acid (HCN). HCN is distilled and absorbed in NaOH solution, and sample is analyzed for CN.
- With standard taper ClearSeal™ grease-free joints and medium frit
- Wheaton-33™ low extractable borosilicate glass
- Components also available separately; see cat. no. 091081
Compliance
For use according to ASTM D 2036
Specifications
USP Type I, ASTM E438 Type I, Class A | |
Absorber jacket, absorber top, cold finger condenser, cold finger jacket, 1000mL distilling flask, and inlet tube, ClearSeal™ grease-free joints and medium frit | |
Cyanide distillation kit |
Ideal for removing interfering substances in quantitative analysis for cyanide | |
Borosilicate Glass |