Learn More
DWK Life Sciences Wheaton™ CryoELITE™ Cryogenic Storage Vials, Externally Threaded
CryoELITE Vial with Loctagon skirt provides stability in freestanding position, and locks into the CryoELITE Benchmate Rack, making it easy to open and close the vial with one hand.
$124.79 - $814.42
Specifications
Diameter (English) Outer | 0.47 in. |
---|---|
Color | Clear |
Diameter (Metric) Outer | 12 mm |
Material | Polypropylene |
Thread Style | External |
Catalog Number | Mfr. No. | Volume (Metric) | Closure Color | Sterility | Marking Spot | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Closure Color | Sterility | Marking Spot | Price | Quantity | ||
02-912-727
|
DWK Life Sciences
W985862 |
1.2 mL | Natural | Sterile | Yes |
Case of 500 Each for $342.92
|
|
||
02-912-733
|
DWK Life Sciences
W985868 |
2 mL | Blue | Sterile | Yes |
Case of 500 Each for $425.11
|
|
||
03-410-056
|
DWK Life Sciences
W985868100 |
2 mL | Blue | Sterile | Yes |
Case of 100 Each for $194.53
|
|
||
02-912-732
|
DWK Life Sciences
W985867 |
2 mL | Green | Sterile | Yes |
Case of 500 Each for $425.11
|
|
||
03-410-055
|
DWK Life Sciences
W985867100 |
2 mL | Green | Sterile | Yes |
Case of 100 Each for $124.79
|
|
||
02-912-737
|
DWK Life Sciences
W985872 |
2 mL | Natural | Sterile | Yes |
Case of 500 Each for $425.11
|
|
||
03-410-050
|
DWK Life Sciences
W985872100 |
2 mL | Natural | Sterile | Yes |
Case of 100 Each for $193.97
|
|
||
02-912-723
|
DWK Life Sciences
W985852 |
2 mL | Natural | Non-sterile | Yes |
Case of 1000 Each for $814.42
|
|
||
02-912-724
|
DWK Life Sciences
W985853 |
2 mL | Natural | Non-sterile | No |
Case of 1000 Each for $633.13
|
|
||
02-912-730
|
DWK Life Sciences
W985865 |
2 mL | Pink | Sterile | Yes |
Case of 500 Each for $425.11
|
|
||
03-410-053
|
DWK Life Sciences
W985865100 |
2 mL | Pink | Sterile | Yes |
Case of 100 Each for $194.53
|
|
||
02-912-729
|
DWK Life Sciences
W985864 |
2 mL | Red | Sterile | Yes |
Case of 500 Each for $425.11
|
|
||
03-410-052
|
DWK Life Sciences
W985864100 |
2 mL | Red | Sterile | Yes |
Case of 100 Each for $194.53
|
|
||
02-912-728
|
DWK Life Sciences
W985863 |
2 mL | White | Sterile | Yes |
Case of 500 Each for $404.47
|
|
||
03-410-051
|
DWK Life Sciences
W985863100 |
2 mL | White | Sterile | Yes |
Case of 100 Each for $194.53
|
|
||
02-912-731
|
DWK Life Sciences
W985866 |
2 mL | Yellow | Sterile | Yes |
Case of 500 Each for $450.47
|
|
||
03-410-054
|
DWK Life Sciences
W985866100 |
2 mL | Yellow | Sterile | Yes |
Case of 100 Each for $194.53
|
|
||
02-912-734
|
DWK Life Sciences
W985869 |
3 mL | Natural | Sterile | Yes |
Case of 500 Each for $509.08
|
|
||
02-912-735
|
DWK Life Sciences
W985870 |
4 mL | Natural | Sterile | Yes |
Case of 500 Each for $547.43
|
|
||
02-912-736
|
DWK Life Sciences
W985871 |
5 mL | Natural | Sterile | Yes |
Case of 500 Each for $545.43
|
|
||
Description
CryoELITE Vial with Loctagon skirt provides stability in freestanding position, and locks into the CryoELITE Benchmate Rack, making it easy to open and close the vial with one hand.
The unique manufacturing design of the CryoELITE vial creates more cap to vial surface contact for an absolute seal. The freestanding vial accommodates an optional 2D Data Matrix Bar Code Bottom Insert (Mfr. No. W985880); which provides a unique identifier for permanent traceability. The bar code bottom insert; which can be purchased separately, easily press fits into the bottom of the vial. The vial bottom format provides unrestricted sample viewing and allows for automated scanning of bar code especially when used with the Wheaton KeepIT-100™ Freezer Box (Mfr. No. W651700 Series). This results in exceptional cap seal performance exceeding DOT standards by an average of 35%, making the CryoELITE vial ideal for the transport of clinical samples. Caps and vials are made from low binding cryogenic, virgin polypropylene.
- Made from cryogenic, radiation grade, virgin polypropylene
- Seven different cap colors, Blue, Pink, Green, Yellow, Red, White, and Natural
- Externally and internally threaded vials
- Design results in pressure resistance performance well in excess of government standards
Specifications
0.47 in. | |
12 mm | |
External | |
Yes | |
Polypropylene | |
Cryogenic Vial |
Clear | |
Polypropylene | |
Freestanding | |
Yes | |
Screw Cap |