Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Caps
Suitable for most sterilization methods
Supplier:  DWK Life Sciences 891088679
 View more versions of this product
Catalog No. 13-710-922
Only null left
Explore available promotions
Description
Description
  • Made from fluoropolymer
  • Material contact surfacesmade from PTFE
  • Colorant and pigment free
  • Available in two DIN Thread sizes: GL 25 and GL 45
Specifications
Specifications
Premium Screw Cap, DIN Thread Size: GL 45
Fluoropolymer
GL 45
Product Suggestions
Product Suggestions