DWK Life Sciences Wheaton™ Glass Sample Vials with Snap Caps
Ideal for collecting, storing, and exhibiting sample in the lab or field, Wheaton™ Glass Sample Vials with Snap Caps contain caps which fit over a special sealing ring to provide effective barrier against moisture.
Supplier:  DWK Life Sciences 225543
 View more versions of this product
Catalog No. 03-335-10A
Explore available promotions
Description
Description
Ideal for collecting, storing, and exhibiting sample in the lab or field, Wheaton™ Glass Sample Vials with Snap Caps contain caps which fit over a special sealing ring to provide effective barrier against moisture.
  • Excellent storage of hygroscopic materials
Specifications
Specifications
Clear
Soda Lime Glass
For collecting, storing and exhibiting sample in the lab or field
LDPE
50 mm
37 mm
30.0 mL
8 dr.
Snap Cap
USP Type III