DWK Life Sciences Wheaton™ Glass Sample Vials with Snap Caps
Ideal for collecting, storing, and exhibiting sample in the lab or field, Wheaton™ Glass Sample Vials with Snap Caps contain caps which fit over a special sealing ring to provide effective barrier against moisture.
Supplier:  DWK Life Sciences 225536
 View more versions of this product
Catalog No. 03-335-20A
Explore available promotions
Description
Description
Ideal for collecting, storing, and exhibiting sample in the lab or field, Wheaton™ Glass Sample Vials with Snap Caps contain caps which fit over a special sealing ring to provide effective barrier against moisture.
  • Excellent storage of hygroscopic materials
Specifications
Specifications
Clear
Soda Lime Glass
For collecting, storing and exhibiting sample in the lab or field
Wheaton
LDPE
40 mm
Vial
30 mm
16.0 mL
4.3 dr.
16 mL
Snap Cap
USP Type III