DWK Life Sciences Wheaton™ Glass Staining Dishes for 50 Slides
Holds up to 50 slides horizontally
Supplier:  DWK Life Sciences 900403
 View more versions of this product
Catalog No. 08-813C
Explore available promotions
Description
Description
  • Dish and cover are made from soda-lime glass
  • Dimensions: 3.25 L × 7.875 W × 4.125 in. H (80 × 200 × 105 mm)
Rack
  • Nontarnishable and withstands most staining solutions
  • Holds up to 50 slides horizontally
  • Hinged handle allows easy insertion and removal of slides
Specifications
Specifications
Yes
Soda Lime Glass
Glass staining dish for 50 slides
50 slides horizontally
7.2 in.
3.4 in.
2.5 in.
6.5 cm
Rectangular
Staining Dish
Cover
18.5 x 8.8 cm (7.2 x 3.4 in.)
185 mm
88 mm
68 mm