missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Glass Staining Dishes for 50 Slides
Holds up to 50 slides horizontally
Supplier: DWK Life Sciences 900404
Description
- Dish and cover are made from soda-lime glass
- Dimensions: 3.25 L × 7.875 W × 4.125 in. H (80 × 200 × 105 mm)
- Nontarnishable and withstands most staining solutions
- Holds up to 50 slides horizontally
- Hinged handle allows easy insertion and removal of slides
Specifications
No | |
Slotted Rack | |
Rack |
Stainless Steel | |
Glass staining dish for 50 slides | |
50 slides horizontally |