missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ LDPE Dispensing Bottles
Excellent for mild and strong buffers, good chemical resistance
$401.88 - $777.12
Specifications
Color | Natural |
---|---|
Material | LDPE |
Autoclavable | Not Autoclavable |
Product Type | Dispensing Bottle |
Closure Size | 20-410 mm |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
03-439-59D
|
DWK Life Sciences
DWK211194 |
60 mL |
Case of 144 Each for $777.12
|
|
|||||
03-439-59E
|
DWK Life Sciences
211195 |
125 mL |
Case of 72 Each for $401.88
|
|
|||||
Description
- Made from low density polyethylene
- Translucent color
- Very flexible with high impact strength
- Raise the spout to dispense contents and depress spout to seal bottle
Specifications
Natural | |
Not Autoclavable | |
20-410 mm |
LDPE | |
Dispensing Bottle | |
Screw Cap |