DWK Life Sciences Wheaton™ Limited-Volume Inserts for E-Z Vials™
Compatible with numerous vials and caps
$283.08 - $411.97
Products 2
Catalog Number Mfr. No. Material Volume (Metric) Includes Price Quantity  
Catalog Number Mfr. No. Material Volume (Metric) Includes Price Quantity  
03-337-52
View Documents Promotion Details
DWK Life Sciences
225350631
Glass 0.35 mL With flat bottom
Case of 1000 Each for $411.97
03-340-149
View Documents Promotion Details
DWK Life Sciences
225265
Glass 0.25 mL With bottom spring
Pack of 100 Each for $283.08
Description
Description
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'