DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps
For use with Wheaton media bottles
Supplier: DWK Life Sciences 240080
Description
- Black phenolic caps with screw thread
- Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)
Specifications
Black | |
200 Cs. | |
Media bottle |
Polyethylene | |
33-430 |