DWK Life Sciences Wheaton™ Prescored Gold-Band™ Amber Ampules
Protect light-sensitive samples
Supplier:  DWK Life Sciences 176796
 View more versions of this product
Catalog No. 01-215P
Explore available promotions
Description
Description
  • Amber borosilicate glass
  • Prescored to eliminate filing
  • Gold-Band feature indicates prescoring
  • Stems can be pull sealed or tip sealed
Compliance
Meet USP Type 1 and ASTM™ Type I, Class B standards
Specifications
Specifications
2 mL
Prescored Ampule
1.2 cm
7.5 cm
USP Type 1, ASTM Type I, Class A
Borosilicate Glass
Amber
0.472 in.
2.95 in.
Videos
SDS
Documents
Documents
Accessories