DWK Life Sciences Wheaton™ Scrubber Absorber Tube
For arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method
Supplier: DWK Life Sciences 180034
Description
- Design eliminates the ball and socket joint connecting the scrubber and absorber components
- Assembled apparatus is freestanding and easy to use
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Applications:
For arsenic analysis
Specifications
Scrubber/Absorber Tube | |
For arsenic analysis using silver diethyldithiocarbamate method | |
Borosilicate Glass | |
1/Cs. |