DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier:  DWK Life Sciences 223760
 View more versions of this product
Catalog No. 06-402A
Explore available promotions
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Specifications
0.17 oz., 5 mL
0.17 oz.
Borosilicate Glass
1.85 in.
5 mL
Amber
47 mm
23 mm