DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier: DWK Life Sciences 223762
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Specifications
0.7 oz., 20 mL | |
0.7 oz. | |
1.25 in. | |
58 mm | |
32 mm |
20 mL | |
Amber | |
Borosilicate Glass | |
2.28 in. |