DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier: DWK Life Sciences 223763
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Specifications
1.01 oz., 30 mL | |
1.01 oz. | |
1.45 in. | |
62 mm | |
37 mm |
30 mL | |
Amber | |
Borosilicate Glass | |
2.44 in. |