DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier:  DWK Life Sciences 223766
 View more versions of this product
Catalog No. 06-402F
Explore available promotions
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Specifications
3.38 oz., 100 mL
3.38 oz.
2.04 in.
95 mm
52 mm
100 mL
Amber
Borosilicate Glass
3.74 in.