DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier: DWK Life Sciences W223712
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Specifications
0.1 oz., 2 mL | |
0.1 oz. | |
0.61 in. | |
36 mm | |
15.6 mm |
2 mL | |
Clear | |
Borosilicate Glass | |
1.41 in. |