DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier:  DWK Life Sciences 223739
 View more versions of this product
Catalog No. 06-406D
Explore available promotions
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Specifications
0.34 oz., 10 mL
0.34 oz.
0.98 in.
54 mm
25 mm
10 mL
Clear
Borosilicate Glass
2.12 in.